MilliporeSigma
  • Inhibition of glycolytic activator PFKFB3 suppresses tumor growth and induces tumor vessel normalization in hepatocellular carcinoma.
Inhibition of glycolytic activator PFKFB3 suppresses tumor growth and induces tumor vessel normalization in hepatocellular carcinoma.
Cancer letters (2020-12-12)
Kenichi Matsumoto, Takehiro Noda, Shogo Kobayashi, Yoshihiro Sakano, Yuki Yokota, Yoshifumi Iwagami, Daisaku Yamada, Yoshito Tomimaru, Hirofumi Akita, Kunihito Gotoh, Yutaka Takeda, Masahiro Tanemura, Koji Umeshita, Yuichiro Doki, Hidetoshi Eguchi
ABSTRACT
Glycolysis emerges as a new therapeutic target for malignancies. The inhibition of glycolytic activator, PFKFB3, repairs tumor endothelial cell function, and normalizing the tumor microenvironment. We aimed to investigate the significance of PFKFB3 in HCC, and the effects of the PFKFB3 inhibitor, PFK15, in HCC tumor cells and tumor endothelial cells. Double immunofluorescent staining of PFKFB3 and CD31 in HCC tissues revealed that high PFKFB3 expression in both tumor cells and tumor endothelial cells was significantly correlated with poor prognosis. Multivariate analysis identified PFKFB3 expression as an independent prognostic factor. PFK15 suppressed proliferation of HCC cell line and tumor endothelial cells in vitro. In a subcutaneous tumor model of the HCC cell line with tumor endothelial cells, PFK15 suppressed tumor growth and induced apoptosis. Moreover, PFK15 treatment induced tumor vessel normalization, decreasing vessel diameter with pericyte attachment and improving vessel perfusion. High PFKFB3 expression in both tumor cells and tumor endothelial cells was identified as a novel prognostic marker in HCC. Targeting PFKFB3 via PFK15 might be a promising strategy for suppressing tumor growth and inducing tumor vessel normalization.