MilliporeSigma
  • Coupling of N-tosylhydrazones with tetrazoles: synthesis of 2-β-D-glycopyranosylmethyl-5-substituted-2H-tetrazole type glycomimetics.
Coupling of N-tosylhydrazones with tetrazoles: synthesis of 2-β-D-glycopyranosylmethyl-5-substituted-2H-tetrazole type glycomimetics.
Organic & biomolecular chemistry (2020-12-24)
Tímea Kaszás, Ivett Cservenyák, Éva Juhász-Tóth, Andrea E Kulcsár, Paola Granatino, Ulf J Nilsson, László Somsák, Marietta Tóth
ABSTRACT
Coupling reactions of O-peracylated 2,6-anhydro-aldose tosylhydrazones (C-(β-d-glycopyranosyl)formaldehyde tosylhydrazones) with tetrazoles were studied under metal-free conditions using thermic or microwave activation in the presence of different bases. The reactions proved highly regioselective and gave the corresponding, up-to-now unknown 2-β-d-glycopyranosylmethyl-2H-tetrazoles in 7-67% yields. The method can be applied to get new types of disaccharide mimetics, 5-glycosyl-2-glycopyranosylmethyl-2H-tetrazoles, as well. Galectin binding studies with C-(β-d-galactopyranosyl)formaldehyde tosylhydrazone and 2-(β-d-galactopyranosylmethyl)-5-phenyl-2H-tetrazole revealed no significant inhibition of any of these lectins.
Sigma-Aldrich
1,3-Propylene sulfite, 99%