MilliporeSigma
  • Site-specific electrodeposition enables self-terminating growth of atomically dispersed metal catalysts.
Site-specific electrodeposition enables self-terminating growth of atomically dispersed metal catalysts.
Nature communications (2020-09-13)
Yi Shi, Wen-Mao Huang, Jian Li, Yue Zhou, Zhong-Qiu Li, Yun-Chao Yin, Xing-Hua Xia
ABSTRACT
The growth of atomically dispersed metal catalysts (ADMCs) remains a great challenge owing to the thermodynamically driven atom aggregation. Here we report a surface-limited electrodeposition technique that uses site-specific substrates for the rapid and room-temperature synthesis of ADMCs. We obtained ADMCs by the underpotential deposition of a non-noble single-atom metal onto the chalcogen atoms of transition metal dichalcogenides and subsequent galvanic displacement with a more-noble single-atom metal. The site-specific electrodeposition enables the formation of energetically favorable metal-support bonds, and then automatically terminates the sequential formation of metallic bonding. The self-terminating effect restricts the metal deposition to the atomic scale. The modulated ADMCs exhibit remarkable activity and stability in the hydrogen evolution reaction compared to state-of-the-art single-atom electrocatalysts. We demonstrate that this methodology could be extended to the synthesis of a variety of ADMCs (Pt, Pd, Rh, Cu, Pb, Bi, and Sn), showing its general scope for functional ADMCs manufacturing in heterogeneous catalysis.
Sigma-Aldrich
Rhodium(III) chloride, 98%
Pricing and availability is not currently available.