MilliporeSigma
  • Challenges in Ki-67 assessments in pulmonary large-cell neuroendocrine carcinomas.
Challenges in Ki-67 assessments in pulmonary large-cell neuroendocrine carcinomas.
Histopathology (2020-10-08)
Ann E Walts, James M Mirocha, Alberto M Marchevsky
ABSTRACT
To gather the best available evidence regarding Ki-67% values in large-cell neuroendocrine carcinoma (LCNEC) and determine whether certain cut-off values could serve as a prognostic feature in LCNEC. Aperio ScanScope AT Turbo, eSlide Manager and ImageScope software (Leica Biosystems) were used to measure Ki-67% in 77 resected LCNEC diagnosed by World Health Organisation (WHO) criteria. Cases were stratified into six classes by 10% Ki-67 increments. Using the Kaplan-Meier method, overall (OS) and disease-free survivals (DFS) were compared by AJCC stage, by six Ki-67% classes and with Ki-67% cut-points ≥20% and ≥40%. Tumours were from 0.9 to 11.5 cm and pathological stages 1-3. The system measured Ki-67% positivity using 4072-44 533 tumour nuclei per case (mean 16610 ± 8039). Ki-67% ranged from 1 to 64% (mean = 26%; median = 26%). Only 16 (21%) tumours had Ki-67% ≥40%. OS ranged from 1 to 298 months (median follow-up = 25 months). DFS ranged from 1 to 276 months (median follow-up = 9 months). OS and DFS differed across AJCC stage (overall log-rank P = 0.038 and P = 0.037). However, neither OS nor DFS significantly correlated with Ki-67% when six or two classes were used with either ≥20% Ki-67 or ≥40% Ki-67 as cut-point. A literature review identified 14 reports meeting our inclusion criteria with ≥10 LCNEC. Reported Ki-67% ranged from 2% to 100%. Problems contributing to variability in Ki-67% measurements are discussed. Our findings caution against a blanket use of 20%, 40% or other Ki-67% cut-points for LCNEC diagnosis or prognostication.