MilliporeSigma
  • YAP promotes the proliferation and migration of colorectal cancer cells through the Glut3/AMPK signaling pathway.
YAP promotes the proliferation and migration of colorectal cancer cells through the Glut3/AMPK signaling pathway.
Oncology letters (2021-03-12)
Linhua Jiang, Jiawen Zhang, Qixuan Xu, Bin Wang, Yizhou Yao, Liang Sun, Xuchao Wang, Diyuan Zhou, Ling Gao, Shiduo Song, Xinguo Zhu
ABSTRACT
Yes-associated protein (YAP), as a major downstream effector in the Hippo signaling pathway, is considered as an oncogene in cancer. The present study aimed to investigate the potential role of YAP in the development and progression of colorectal cancer (CRC). The mRNA and protein expression levels of YAP in human CRC tissue samples and adjacent normal tissue were analyzed using public databases, as well as clinical samples. The potential roles of YAP and the underlying mechanism regulating the proliferation and migration of CRC cells were examined using genetic manipulation in vitro. The correlation between the expression of the YAP gene and epithelial-to-mesenchymal transition (EMT) markers was investigated in order to determine the mechanism underlying the observed effects of YAP. YAP mRNA expression levels were significantly upregulated in CRC tissue compared with in normal tissue, as determined using datasets obtained from Oncomine. Similarly, in clinical samples, the protein expression levels of YAP were significantly upregulated in CRC tissue samples compared with in normal tissue samples. YAP knockdown inhibited the proliferation and migration of CRC cells in vitro, whereas its overexpression resulted in the opposite effect. The expression levels of the YAP gene were positively correlated with those of EMT markers (such as vimentin and N-cadherin) and EMT-inducing transcription factors (such as Snail1, Slug and zinc finger E-box binding homeobox 1 and 2) in CRC samples from Gene Expression Profiling Interactive Analysis. Furthermore, YAP silencing increased the protein expression of E-cadherin and decreased that of vimentin in CRC cells. By contrast, the overexpression of YAP had the opposite effect. YAP promoted the glucose transporter 3 (Glut3)/AMP-activated protein kinase (AMPK) signaling pathway in CRC cells. In conclusion, YAP promoted the proliferation and migration of CRC cells, as well as the expression of EMT markers, possibly by regulating the Glut3/AMPK signaling pathway.
Sigma-Aldrich
MISSION® esiRNA, targeting human SLC2A3