MilliporeSigma
  • Ecological risk assessment of persistent organic pollutants (POPs) in surface sediments from aquaculture system.
Ecological risk assessment of persistent organic pollutants (POPs) in surface sediments from aquaculture system.
Chemosphere (2020-12-11)
Nuning Vita Hidayati, Laurence Asia, Imen Khabouchi, Franck Torre, Ita Widowati, Agus Sabdono, Pierre Doumenq, Agung Dhamar Syakti
ABSTRACT
Organochlorinated pesticides (OCPs) and Polychlorinated biphenyls (PCBs) in the surface sediments from shrimp ponds in four regions of the northern part of the Central Java coast (namely Brebes, Tegal, Pemalang, and Pekalongan) were investigated. The highest concentration of ∑ OCPs was found in Brebes Regency, ranging from 68.1 ± 3.4 to 168.1 ± 9.8 μg kg-1 dw. As indicated by the DDT ratio and chlordane ratio, the value suggested that those compounds may mainly originate from historical inputs rather than a recent application. The concentrations of Ʃ 7 indicator PCBs were determined, with the concentration ranged from 1.2 ± 0.7 μg kg-1 dw (Pekalongan) to 2.2 ± 0.4 μg kg-1 dw (Tegal). The most toxic PCB congener, PCB 118, was detected in all studied regions, with the highest proportion found in Tegal. Source analysis indicated that PCBs in the sediments mainly originated from Aroclor 1254 and Aroclor 1248. Compared to sediment quality guidelines (SQGs), some OCPs were found with concentrations which potentially posed an adverse effect. Our findings suggested that more attention should be paid to ensure sustainable shrimp culture facing such a risk of the OCPs and PCBs.