MilliporeSigma
  • Terazosin reduces steroidogenic factor 1 and upregulates heat shock protein 90 expression in LH-induced bovine ovarian theca cells.
Terazosin reduces steroidogenic factor 1 and upregulates heat shock protein 90 expression in LH-induced bovine ovarian theca cells.
Free radical biology & medicine (2020-12-23)
Zi-Run Tang, Shou-Long Deng, Zheng-Xing Lian, Kun Yu
ABSTRACT
Hyperthecosis syndrome is a common endocrine system metabolic disorder in women of childbearing age. The main symptoms are elevated androgen levels, abnormal ovulation, and excessive oxidative stress. Currently, there is no effective treatment for hyperthecosis syndrome. α(1)-adrenergic receptor (ADRA1) is involved in the metabolic pathway of ovarian steroid hormone. This study studied the mechanism of the ADRA1 inhibitor terazosin in the LH-induced bovine theca cells in vitro. We found that terazosin regulates the expression of steroidogenic factor 1 (SF1) and downstream genes through the ERK1/2 pathway, reducing androgen content. Terazosin promotes the expression of HSP90 and reduces the activity of iNOS. In addition, Terazosin up-regulates the expression of nuclear factor erythroid 2-related factor 2 (Nrf2) and its downstream gene γ-GCS, which improves the ability of theca cells to resist oxidative stress. This study provides a reference for the treatment of human hyperthecosis syndrome.
Sigma-Aldrich
Terazosin hydrochloride, ≥98% (TLC), powder