MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Non-covalent inhibitors of rhinovirus 3C protease.
Bioorganic & medicinal chemistry letters (2010-12-25)
Andrew Baxter, Mark Chambers, Fredrik Edfeldt, Karl Edman, Adrian Freeman, Cristian Johansson, Sarah King, Andy Morley, Jens Petersen, Phil Rawlins, Loredana Spadola, Bob Thong, Hervé Van de Poël, Nicola Williams
ABSTRACT
The first known non-covalent inhibitors of rhinovirus 3C protease (3CP) have been identified through fragment based screening and hit identification activities.
Sigma-Aldrich
Flavone, ≥99.0%