MilliporeSigma
  • Non-nutritive sweeteners are in concomitant with the formation of endogenous and exogenous advanced glycation end-products.
Non-nutritive sweeteners are in concomitant with the formation of endogenous and exogenous advanced glycation end-products.
International journal of food sciences and nutrition (2020-01-11)
Permal Deo, Christine Chern, Brock Peake, Sze-Yen Tan
ABSTRACT
This study investigated the role of non-nutritive sweeteners in the formation of advanced glycation end-products (AGEs) and their reactive intermediates using endogenous and exogenous models. In the endogenous model, xylitol and sorbitol formed similar levels of reactive intermediates compared to sucralose. Protein-bound fluorescent AGEs, Nε-carboxymethyllysine (CML), and Nε-carboxyethyllysine (CEL) levels in the xylitol and sorbitol treatment were significantly higher compared to the sucralose treatment. In the exogenous model, sucralose treatment showed significantly higher glyoxal and fructosamine levels compared to xylitol and sorbitol, respectively. However, protein-bound fluorescent AGEs, CML, and CEL were lower in the sucralose treatment compared to other sugar treatments. The data suggest that the structure of sugar alcohols which are similar to reducing sugars may contribute to the formation of AGEs and their reactive intermediates in the endogenous model. The long-term effects of non-nutritive sweeteners consumption on AGEs formation and health implications should be verified with population studies.
Sigma-Aldrich
1-Deoxy-1-morpholino-D-fructose, ≥98% (TLC)