MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Demonstration of flat-top beam illumination in widefield multiphoton microscopy.
Demonstration of flat-top beam illumination in widefield multiphoton microscopy.
Journal of biomedical optics (2019-11-16)
Mohammad Kabir, Hemangg Rajput, Varun Kelkar, Adriana Salazar Coariti, Kimani Toussaint
ABSTRACT
Multiphoton microscopy provides a suitable technique for imaging biological tissues with submicrometer resolution. Usually a Gaussian beam (GB) is used for illumination, leading to a reduced power efficiency in the multiphoton response and vignetting for a square-shaped imaging area. A flat-top beam (FTB) provides a uniform spatial intensity distribution that equalizes the probability of a multiphoton effect across the imaging area. We employ a customized widefield multiphoton microscope to compare the performance of a square-shaped FTB illumination with that based on using a GB, for both two-photon fluorescence (TPF) and second-harmonic generation (SHG) imaging. The variation in signal-to-noise ratio across TPF images of fluorescent dyes spans ∼5.6  dB for the GB and ∼1.2  dB for the FTB illumination, respectively. For the GB modality, TPF images of mouse colon and Convallaria root, and SHG images of chicken tendon and human breast biopsy tissue showcase ∼20  %   area that are not imaged due to either insufficient or lack of illumination. For quantitative analysis that depends on the illuminated area, this effect can potentially lead to inaccuracies. This work emphasizes the applicability of FTB illumination to multiphoton applications.