MilliporeSigma
Sensing soluble uric acid by Naip1-Nlrp3 platform.
Cell death & disease (2021-02-07)
Tarcio Teodoro Braga, Mariana Rodrigues Davanso, Davi Mendes, Tiago Antonio de Souza, Anderson Fernandes de Brito, Mario Costa Cruz, Meire Ioshie Hiyane, Dhemerson Souza de Lima, Vinicius Nunes, Juliana de Fátima Giarola, Denio Emanuel Pires Souto, Tomasz Próchnicki, Mario Lauterbach, Stellee Marcela Petris Biscaia, Rilton Alves de Freitas, Rui Curi, Alessandra Pontillo, Eicke Latz, Niels Olsen Saraiva Camara
ABSTRACT
Uric acid (UA), a product of purine nucleotide degradation able to initiate an immune response, represents a breakpoint in the evolutionary history of humans, when uricase, the enzyme required for UA cleavage, was lost. Despite being inert in human cells, UA in its soluble form (sUA) can increase the level of interleukin-1β (IL-1β) in murine macrophages. We, therefore, hypothesized that the recognition of sUA is achieved by the Naip1-Nlrp3 inflammasome platform. Through structural modelling predictions and transcriptome and functional analyses, we found that murine Naip1 expression in human macrophages induces IL-1β expression, fatty acid production and an inflammation-related response upon sUA stimulation, a process reversed by the pharmacological and genetic inhibition of Nlrp3. Moreover, molecular interaction experiments showed that Naip1 directly recognizes sUA. Accordingly, Naip may be the sUA receptor lost through the human evolutionary process, and a better understanding of its recognition may lead to novel anti-hyperuricaemia therapies.
Sigma-Aldrich
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
C75, ≥98% (HPLC), powder
Sigma-Aldrich
TOFA, ≥98% (HPLC)
Sigma-Aldrich
BMS-303141, ≥98% (HPLC)
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder