MilliporeSigma
  • Polymorphism -509C/T in TGFB1 Promoter Is Associated With Increased Risk and Severity of Persistent Allergic Rhinitis in a Chinese Population.
Polymorphism -509C/T in TGFB1 Promoter Is Associated With Increased Risk and Severity of Persistent Allergic Rhinitis in a Chinese Population.
American journal of rhinology & allergy (2020-03-29)
Xin-Jie Zhu, Mei-Ping Lu, Ruo-Xi Chen, Dong-Yun Bu, Lu-Ping Zhu, Mei-Lin Wang, Min Yin, Zheng-Dong Zhang, Lei Cheng
ABSTRACT
Polymorphism -509C/T in the promoter of transforming growth factor beta1 (TGFB1) gene is implicated in the pathogenesis of asthma. This polymorphism might also act to regulate the development of allergic rhinitis (AR). To investigate whether -509C/T is associated with AR susceptibility and severity in a Han Chinese population. The study enrolled 263 patients with persistent AR and 249 healthy controls. AR patients were classified as mild or moderate/severe AR groups according to the Allergic Rhinitis and its Impact on Asthma classification. TGFB1 gene polymorphism -509C/T was genotyped with polymerase chain reaction-restriction fragment length polymorphism (PCR-RFLP). Serum total Immunoglobulin E (IgE) and specific IgE levels were determined using an ImmunoCAP. Significant difference was found in the allele frequency of TGFB1 -509C/T between AR patients and healthy controls (P = .027) but not in the genotype frequency (P =.051). However, the genotype frequency of TGFB1 -509C/T showed significant difference between the mild AR group, the moderate/severe AR group, and the control group (P = .012); between the moderate/severe AR group and the control group (P =.036); between the mild AR group and the moderate/severe AR group (P = .038); but not between the mild AR group and the control group (P =.075). TGFB1 promoter polymorphism -509C/T may be associated with the susceptibility and the severity of persistent AR of Han Chinese, but the functional relationship still needs clarification.