MilliporeSigma
  • Physicochemical characteristics and resistant starch of different varieties of banana from Taiwan
Physicochemical characteristics and resistant starch of different varieties of banana from Taiwan
International journal of food properties (2020)
Li M C, et al.