MilliporeSigma
  • A systems-level study reveals host-targeted repurposable drugs against SARS-CoV-2 infection.
A systems-level study reveals host-targeted repurposable drugs against SARS-CoV-2 infection.
Molecular systems biology (2021-08-03)
Fangyuan Chen, Qingya Shi, Fen Pei, Andreas Vogt, Rebecca A Porritt, Gustavo Garcia, Angela C Gomez, Mary Hongying Cheng, Mark E Schurdak, Bing Liu, Stephen Y Chan, Vaithilingaraja Arumugaswami, Andrew M Stern, D Lansing Taylor, Moshe Arditi, Ivet Bahar
ABSTRACT
Understanding the mechanism of SARS-CoV-2 infection and identifying potential therapeutics are global imperatives. Using a quantitative systems pharmacology approach, we identified a set of repurposable and investigational drugs as potential therapeutics against COVID-19. These were deduced from the gene expression signature of SARS-CoV-2-infected A549 cells screened against Connectivity Map and prioritized by network proximity analysis with respect to disease modules in the viral-host interactome. We also identified immuno-modulating compounds aiming at suppressing hyperinflammatory responses in severe COVID-19 patients, based on the transcriptome of ACE2-overexpressing A549 cells. Experiments with Vero-E6 cells infected by SARS-CoV-2, as well as independent syncytia formation assays for probing ACE2/SARS-CoV-2 spike protein-mediated cell fusion using HEK293T and Calu-3 cells, showed that several predicted compounds had inhibitory activities. Among them, salmeterol, rottlerin, and mTOR inhibitors exhibited antiviral activities in Vero-E6 cells; imipramine, linsitinib, hexylresorcinol, ezetimibe, and brompheniramine impaired viral entry. These novel findings provide new paths for broadening the repertoire of compounds pursued as therapeutics against COVID-19.
Sigma-Aldrich
Furin Inhibitor I, Furin Inhibitor I, is a peptidyl chloromethylketone that binds to the catalytic site of furin and blocks its activity.