MilliporeSigma
  • Synthesis and evaluation of anti-tubercular and antibacterial activities of new 4-(2,6-dichlorobenzyloxy)phenyl thiazole, oxazole and imidazole derivatives. Part 2.
Synthesis and evaluation of anti-tubercular and antibacterial activities of new 4-(2,6-dichlorobenzyloxy)phenyl thiazole, oxazole and imidazole derivatives. Part 2.
European journal of medicinal chemistry (2012-01-24)
Xiaoyun Lu, Xiaobo Liu, Baojie Wan, Scott G Franzblau, Lili Chen, Changlin Zhou, Qidong You
ABSTRACT
A series of substituted 4-(2,6-dichlorobenzyloxy)phenyl thiazole, oxazole and imidazole derivatives were synthesized. The derivatives were screened for in vitro anti-tubercular activities against Mycobacterium tuberculosis H37Rv using the Microplate Alamar Blue Assay (MABA), and antibacterial activities with agar dilution method against clinical S. aureus, E. coli, S. pneumoniae and penicilin-resistant S. pneumoniae. Among 15 compounds, several thiazole derivatives exhibited good anti-tubercular activities with MIC values between 1 μM and 61.2 μM, and potent activities against S. pneumoniae with MIC values less than 0.134 μM. These studies suggest that the thiazole scaffold may serve as a new promising template for further elaboration as anti-tubercular and antibacterial drugs.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Supelco
Isoniazid, analytical standard, ≥99% (TLC)