MilliporeSigma
  • Conjugated linoleic acid conversion by six Lactobacillus plantarum strains cultured in MRS broth supplemented with sunflower oil and soymilk.
Conjugated linoleic acid conversion by six Lactobacillus plantarum strains cultured in MRS broth supplemented with sunflower oil and soymilk.
Journal of Food Science (2017)
Li, et al.
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid