MilliporeSigma
Nucleoporin genes in human diseases.
European journal of human genetics : EJHG (2016-04-14)
Valeria Nofrini, Danika Di Giacomo, Cristina Mecucci
ABSTRACT
Nuclear pore complexes (NPCs) are large channels spanning the nuclear envelope that mediate nucleocytoplasmic transport. They are composed of multiple copies of ~30 proteins termed nucleoporins (NUPs). Alterations in NUP genes are linked to several human neoplastic and non-neoplastic diseases. This review focuses on NUPs, their genes, localization, function in the NPC and involvement in human diseases.