MilliporeSigma
  • An alternative, effective substrate for erythrocyte acid phosphatase phenotype determinations.
An alternative, effective substrate for erythrocyte acid phosphatase phenotype determinations.
Journal of forensic sciences (1988-07-01)
B Budowle, A M Gambel
ABSTRACT
A method is described for obtaining nondiffusing, nonfading fluorogenic zymograms for erythrocyte acid phosphatase variants separated by isoelectric focusing. The synthetic substrate 4-trifluoromethylcoumarin phosphate was impregnated into cellulose diacetate membranes and air-dried overnight. After isoelectric focusing, the substrate overlay membrane was rehydrated in 0.05M citrate buffer, pH 4.0, lightly blotted, and overlayed on the gel. A 5- to 10-min incubation at 37 degrees C produced intensely fluorescing, light-blue bands on the diacetate membrane. Interaction of the trifluoromethyl group on the substrate with hydrophobic regions of the diacetate membrane impeded diffusion, yielding a permanent zymogram. There were no discrepancies in phenotype determinations using this method when compared with the 4-methylumbelliferyl phosphate assay approach. Further, an increased number of conclusive calls were obtained (91.8 versus 79.5% and 54.1 versus 34.9%) with this new assay when compared with the 4-methylumbelliferyl phosphate substrate on known liquid bloods and questioned dried bloodstains, respectively.