MilliporeSigma
  • Turbidimetric measurement of $\gamma$-globulin in cerebrospinal fluid with use of sulfosalicylic acid and Triton X-100
Turbidimetric measurement of $\gamma$-globulin in cerebrospinal fluid with use of sulfosalicylic acid and Triton X-100
Journal of Analytical Bio-Science Vol (2009)
Ihara H, et al.