MilliporeSigma
  • Control of Mozzarella spoilage bacteria by using bovine lactoferrin pepsin-digested hydrolysate
Control of Mozzarella spoilage bacteria by using bovine lactoferrin pepsin-digested hydrolysate
Worldwide Research Efforts in the Fighting Against Microbial Pathogens From Basic Research to Technological Developments (2013)