MilliporeSigma
  • Genetic variants of bovine $\beta$-and $\kappa$-casein result in different immunoglobulin E-binding epitopes after in vitro gastrointestinal digestion
Genetic variants of bovine $\beta$-and $\kappa$-casein result in different immunoglobulin E-binding epitopes after in vitro gastrointestinal digestion
Journal of Dairy Science (2013)
Lisson M, et al.