MilliporeSigma
  • Asymmetric superwetting stainless steel meshes for on-demand and highly effective oil-water emulsion separation
Asymmetric superwetting stainless steel meshes for on-demand and highly effective oil-water emulsion separation
Separation and Purification Technology (2021)
Qiu L, et al.