MilliporeSigma
  • A robust copper oxide-based superhydrophobic microfiltration membrane for moisture-proof treatment of trace water in transformer oil
A robust copper oxide-based superhydrophobic microfiltration membrane for moisture-proof treatment of trace water in transformer oil
Colloids and Surfaces. A, Physicochemical and Engineering Aspects (2021)
Zhang J, et al.