MilliporeSigma
  • A Proteolytic Enzyme of Streptomyces griseus: I. Purification of a Protease of Streptomyces griseus
A Proteolytic Enzyme of <I>Streptomyces griseus</I>: I. Purification of a Protease of <I>Streptomyces griseus</I>
Biochemistry (1959)
Nomoto, M. and Y. Narahashi