MilliporeSigma
  • Developmental phase-specific alkaline phosphatase isoenzymes of human placenta and their occurrence in human cancer.
Developmental phase-specific alkaline phosphatase isoenzymes of human placenta and their occurrence in human cancer.
Cancer research (1976-07-01)
L Fishman, H Miyayama, S G Driscoll, W H Fishman
ABSTRACT
Alkaline phosphatase electrophoretic patterns characteristic of three phases in early human trophoblast development are described in this preliminary communication. Phase 1 (6 to 10 weeks) consists entirely of two heat-sensitive, L-homoarginine-inhibited bands, the slower one of which possesses antigenic determinants of live-bone-type alkaline phosphatase, whereas the fast band lacks any of the known alkaline phosphatase antigenic determinants. Phase 2 pattern (11 to 13 weeks) is that of a mixture of Phase 1 and Phase 3 isozyme components, the latter exhibiting two isozyme bands with the characteristics of term placental alkaline phosphatases correspond in order to non-Regan isoenzyme, a mixture of Regan and non-Regan isoenzymes and Regan isoenzyme in a variety of human cancer tissues. The biochemical profile characteristic of trophoblast developmental Phase 1 alkaline phosphatase is expressed as 78.5% heat-sensitive inhibition (5 min at 65 degrees), 66.3% L-homoarginine inhibition, and 17.3% L-phenylalanine inhibition where n = 12. It is hypothesized that the alkaline phosphatase of human tumor tissues reflects the expression of placental genes corresponding to one or more phases of trophoblastic development.