MilliporeSigma
  • Sputum and Plasma Neutrophil Elastase in Stable Adult Patients With Cystic Fibrosis in Relation to Chronic Pseudomonas Aeruginosa Colonization.
Sputum and Plasma Neutrophil Elastase in Stable Adult Patients With Cystic Fibrosis in Relation to Chronic Pseudomonas Aeruginosa Colonization.
Cureus (2021-07-06)
Atqah AbdulWahab, Mona Allangawi, Merlin Thomas, Ilham Bettahi, Siveen K Sivaraman, Jayakumar Jerobin, Prem Chandra, Manjunath Ramanjaneya, Abdul-Badi Abou-Samra
ABSTRACT
Background and purpose Neutrophil elastase (NE) has been implicated in the pathogenesis of airway inflammation in cystic fibrosis (CF) patients and it impairs defenses against Pseudomonas aeruginosa (PA) infection or colonization. Sputum NE may act as a biomarker of neutrophilic inflammation in CF patients. This study aimed to determine sputum and plasma total NE levels in clinically stable adult CF patients and control subjects, and their correlation to PA colonization and lung functions. Methods This is a cross-sectional study. Total NE was measured on spontaneously expectorated sputum and plasma obtained from 21 CF patients, aged 18-40 years, during routine visits to the adult CF clinic. This was compared to plasma obtained from 22 matching healthy controls. The levels of NE were measured by the magnetic bead-based multiplex assay. Results Sputum and plasma NE levels had a significant positive correlation (Pearson r=0.533, P=0.013) with PA colonization. Sixteen CF patients (76.2%) were chronically colonized with PA. Both median sputum and plasma NE were found to be higher in CF patients with PA as compared with non-PA patients, even though this difference was statistically insignificant. Sputum and plasma NE levels did not correlate with the percentage predicted forced expiratory volume in one second (FEV1), the forced vital capacity (FVC), and FEV1/FVC and no association with PA. Conclusion The findings suggest that clinically stable adult CF patients colonized with PA may have higher NE levels in both plasma and sputum as compared to non-PA CF patients and probably total NE does not influence lung functions.
Product Description
Millipore
MILLIPLEX® Human Sepsis Panel