MilliporeSigma
  • An unstable manganese(III) complex incorporating ligand donor types proposed for an acid phosphatase from sweet potato: (p-nitrobenzenethiolato)[N,N'-ethylenebis(salicylideneaminato)]manganese(III)
An unstable manganese(III) complex incorporating ligand donor types proposed for an acid phosphatase from sweet potato: (p-nitrobenzenethiolato)[N,N'-ethylenebis(salicylideneaminato)]manganese(III)
Inorganic Chemistry (1988)
Gohdes, J. and W. Armstrong