MilliporeSigma
  • Nerve growth factor variations in patients with mood disorders: no changes in eight weeks of clinical treatment.
Nerve growth factor variations in patients with mood disorders: no changes in eight weeks of clinical treatment.
Neuropsychiatric disease and treatment (2014-05-29)
Xiaohua Liu, Tianhong Zhang, Shen He, Bo Hong, Daihui Peng, Hui Su, Fei Li, Yingying Tang, Zhiguang Lin, Yiru Fang, Kaida Jiang
ABSTRACT
Nerve growth factor (NGF) has received much attention for its role in mood disorders. The primary objective of the present study was to examine serum NGF levels in Chinese inpatients with depressive or manic episodes in the acute phase and to explore the changes in NGF levels after effective clinical treatments. One hundred and seven consecutive inpatients and outpatients with mood disorders (30 with unipolar depression, 23 with bipolar depression, and 54 with bipolar mania), and 50 healthy controls were recruited. The serum NGF levels were detected by enzyme-linked immunosorbent assay. Patients with bipolar mania presented higher serum NGF levels compared to those of healthy controls. After 8 weeks of medical treatment, there were significant improvements in symptoms in patients, but no significant changes in NGF levels. The present findings may help to strengthen and expand the understanding of the role of NGF in the acute stages of mood disorders.