MilliporeSigma
  • Synthetic Siglec-9 Agonists Inhibit Neutrophil Activation Associated with COVID-19.
Synthetic Siglec-9 Agonists Inhibit Neutrophil Activation Associated with COVID-19.
ChemRxiv : the preprint server for chemistry (2021-01-21)
Corleone S Delaveris, Aaron J Wilk, Nicholas M Riley, Jessica C Stark, Samuel S Yang, Angela J Rogers, Thanmayi Ranganath, Kari C Nadeau, Catherine A Blish, Carolyn R Bertozzi
ABSTRACT
Severe cases of coronavirus disease 2019 (COVID-19), caused by infection with SARS-Cov-2, are characterized by a hyperinflammatory immune response that leads to numerous complications. Production of proinflammatory neutrophil extracellular traps (NETs) has been suggested to be a key factor in inducing a hyperinflammatory signaling cascade, allegedly causing both pulmonary tissue damage and peripheral inflammation. Accordingly, therapeutic blockage of neutrophil activation and NETosis, the cell death pathway accompanying NET formation, could limit respiratory damage and death from severe COVID-19. Here, we demonstrate that synthetic glycopolymers that activate the neutrophil checkpoint receptor Siglec-9 suppress NETosis induced by agonists of viral toll-like receptors (TLRs) and plasma from patients with severe COVID-19. Thus, Siglec-9 agonism is a promising therapeutic strategy to curb neutrophilic hyperinflammation in COVID-19.<br>.
Sigma-Aldrich
SHP1/2 PTPase Inhibitor, NSC-87877, The SHP1/2 PTPase Inhibitor, NSC-87877, also referenced under CAS 56932-43-5, controls the biological activity of SHP1/2 PTPase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Pricing and availability is not currently available.