MilliporeSigma
  • Purification of pro- and eukaryotic superoxide dismutases by charge-controlled hydrophobic chromatography
Purification of pro- and eukaryotic superoxide dismutases by charge-controlled hydrophobic chromatography
Journal of Chromatography A (1992)
Grunow, M. and W. Schopp