MilliporeSigma
  • Loss of Gene Information: Discrepancies between RNA Sequencing, cDNA Microarray, and qRT-PCR.
Loss of Gene Information: Discrepancies between RNA Sequencing, cDNA Microarray, and qRT-PCR.
International journal of molecular sciences (2021-09-11)
Nicole Rachinger, Stefan Fischer, Ines Böhme, Lisa Linck-Paulus, Silke Kuphal, Melanie Kappelmann-Fenzl, Anja K Bosserhoff
ABSTRACT
Molecular analyses of normal and diseased cells give insight into changes in gene expression and help in understanding the background of pathophysiological processes. Years after cDNA microarrays were established in research, RNA sequencing (RNA-seq) became a key method of quantitatively measuring the transcriptome. In this study, we compared the detection of genes by each of the transcriptome analysis methods: cDNA array, quantitative RT-PCR, and RNA-seq. As expected, we found differences in the gene expression profiles of the aforementioned techniques. Here, we present selected genes that exemplarily demonstrate the observed differences and calculations to reveal that a strong RNA secondary structure, as well as sample preparation, can affect RNA-seq. In summary, this study addresses an important issue with a strong impact on gene expression analysis in general. Therefore, we suggest that these findings need to be considered when dealing with data from transcriptome analyses.
Sigma-Aldrich
Monoclonal Anti-SOX21 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL4688, purified immunoglobulin