MilliporeSigma
  • Molecular cloning and nucleotide sequence of the gene for pyruvate kinase of Bacillus stearothermophilus and the production of the enzyme in Escherichia coli. Evidence that the genes for phosphofructokinase and pyruvate kinase constitute an operon
Molecular cloning and nucleotide sequence of the gene for pyruvate kinase of <I>Bacillus stearothermophilus</I> and the production of the enzyme in <I>Escherichia coli</I>. Evidence that the genes for phosphofructokinase and pyruvate kinase constitute an operon
FEBS Journal (1993)
Sakai, H. and T. Ohta