MilliporeSigma
  • Synthesis and pharmacology of the enantiomers of UH301: opposing interactions with 5-HT1A receptors.
Synthesis and pharmacology of the enantiomers of UH301: opposing interactions with 5-HT1A receptors.
Chirality (1996-01-01)
S E Hillver, L Björk, B B Höök, L Cortizo, G Nordvall, A M Johansson, A Ertan, I Csöregh, L Johansson, T Lewander, U Hacksell
ABSTRACT
The (S)-enantiomer of 5-fluoro-8-hydroxy-2-(dipropylamino) tetralin [(S)-2a; (S)-UH301] was the first reported 5-HT1A receptor antagonist. We now give a full account on the synthetic effort leading to the preparation of the racemate and the enantiomers of 2a. The crystal and molecular structure of 2a. HBr has been determined by X-ray diffraction and the absolute configuration has been deduced using statistical tests of the crystallographic R values. The unit cell is tetragonal (P4(1)2(1)2) with a = b = 13.2235(2), c = 39.560(1) A and contains two crystallographically independent molecules in each asymmetric unit. The two solid state conformers differ in the conformation of the N-propyl groups. The pharmacological characterization of the enantiomers was done by use of in vivo biochemical and behavioural assays in rats. The (R)-enantiomer of 2a is a 5-HT1A receptor agonist of low potency while (S)-2a does not exhibit any agonist properties at 5-HT1A receptors. As a consequence of the opposing effects of the enantiomers, the racemate, rac-2a, does not produce any clear-cut effects in rats. The reduced efficacy of (S)-2a as compared to the well known 5-HT1A receptor agonist 8-hydroxy-2-(dipropylamino) tetralin (1;8-OH-DPAT) may be due to the fluoro-substituent induced negative potential of the aromatic ring.