MilliporeSigma
Protocol for chemically induced murine gastric tumor model.
STAR protocols (2021-09-30)
Ke Li, Ao Wang, Huijuan Liu, Baojie Li
ABSTRACT
N-Methyl-N-nitrosourea, an N-nitroso compound converted from dietary nitrite by Helicobacter pylori, causes somatic mutations in epithelial cells and induces gastric premalignancy. Here, we describe a detailed protocol for induction of gastric tumor and analysis of tumor phenotypes in mice. This model can be widely used for studying the initiation and growth of gastric cancer. For complete details on the use and execution of this protocol, please refer to Li et al. (2021).
Sigma-Aldrich
2,2,2-Tribromoethanol, 97%
Sigma-Aldrich
Tamoxifen, ≥99%
Supelco
Ketamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Proteinase K from Tritirachium album