MilliporeSigma
  • A breakthrough-like effect of metformin reduces peripheral resistance to triiodothyronine in euthyroid, non-insulin-resistant, type 2 diabetic patients.
A breakthrough-like effect of metformin reduces peripheral resistance to triiodothyronine in euthyroid, non-insulin-resistant, type 2 diabetic patients.
Endocrine connections (2021-06-18)
Melinda Kertész, Szilárd Kun, Eszter Sélley, Zsuzsanna Nagy, Tamás Kőszegi, István Wittmann
ABSTRACT
Type 2 diabetes is characterized, beyond the insulin resistance, by polyhormonal resistance. Thyroid hormonal resistance has not yet been described in this population of patients. Metformin is used to decrease insulin resistance, and at present, it is assumed to influence the effect of triiodothyronine, as well. In this open-label, pilot, hypothesis-generating, follow-up study, 21 patients were included; all of them were euthyroid with drug naïve, newly diagnosed type 2 diabetes. Before and after 4 weeks of metformin therapy, fructosamine, homeostasis model assessment for insulin resistance (HOMA-IR), thyroid hormones, T3/T4 ratio, and TSH, as well as blood pressure and heart rate using ambulatory blood pressure monitor were measured. We also conducted an in vitro study to investigate the possible mechanisms of T3 resistance, assessing T3-induced Akt phosphorylation among normal (5 mM) and high (25 mM) glucose levels with or without metformin treatment in a human embryonal kidney cell line. Metformin decreased the level of T3 (P < 0.001), the ratio of T3/T4 (P = 0.038), fructosamine (P = 0.008) and HOMA-IR (P = 0.022). All these changes were accompanied by an unchanged TSH, T4, triglyceride, plasma glucose, bodyweight, blood pressure, and heart rate. In our in vitro study, T3-induced Akt phosphorylation decreased in cells grown in 25 mM glucose medium compared to those in 5 mM. Metformin could not reverse this effect. Metformin seems to improve T3 sensitivity in the cardiovascular system in euthyroid, type 2 diabetic patients, the mechanism of which may be supracellular.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, L-glutamine, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture