MilliporeSigma
  • Isolation of the intermediate filament protein vimentin by chromatofocusing.
Isolation of the intermediate filament protein vimentin by chromatofocusing.
FEBS letters (1985-01-28)
H Bloemendal, M Willemsen, G Groenewoud, P Oomen
ABSTRACT
A novel, simple and relatively rapid method is described for the isolation of the intermediate-sized filament protein vimentin from eye lens tissue. Chromatofocusing is applied as the sole purification step. The apparent isoelectric point of the protein in 6 M urea and at 22 degrees C is 4.9. Electrophoretic mobility on one- and two-dimensional polyacrylamide gels, solubility in 6 M urea and amino acid composition were used for identification.