MilliporeSigma
  • Reconstitution of an intact clock reveals mechanisms of circadian timekeeping.
Reconstitution of an intact clock reveals mechanisms of circadian timekeeping.
Science (New York, N.Y.) (2021-10-08)
Archana G Chavan, Jeffrey A Swan, Joel Heisler, Cigdem Sancar, Dustin C Ernst, Mingxu Fang, Joseph G Palacios, Rebecca K Spangler, Clive R Bagshaw, Sarvind Tripathi, Priya Crosby, Susan S Golden, Carrie L Partch, Andy LiWang
ABSTRACT
[Figure: see text].