MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Infectious Subviral Particle to Membrane Penetration Active Particle (ISVP-to-ISVP*) Conversion Assay for Mammalian Orthoreovirus.
Infectious Subviral Particle to Membrane Penetration Active Particle (ISVP-to-ISVP*) Conversion Assay for Mammalian Orthoreovirus.
Bio-protocol (2018-02-06)
Anthony J Snyder, Pranav Danthi
ABSTRACT
The mammalian orthoreovirus (reovirus) outer capsid undergoes a series of conformational changes prior to or during viral entry. These transitions are necessary for delivering the genome-containing core across host cell membranes. This protocol describes an in vitro assay for monitoring the transition into a membrane penetration-active form (i.e., ISVP*).
Product Description
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%