MilliporeSigma
  • Crystallization and preliminary X-ray analysis of pyruvate kinase from Bacillus stearothermophilus
Crystallization and preliminary X-ray analysis of pyruvate kinase from <I>Bacillus stearothermophilus</I>
Acta Crystallographica Section B, Structural Crystallography and Crystal Chemistry (2005)
Suzuki, K.