MilliporeSigma
  • Zirconium (IV) Chloride-Mediated Chemoselective Conjugate Addition of Aliphatic Amines to α, β-Ethylenic Compounds
Zirconium (IV) Chloride-Mediated Chemoselective Conjugate Addition of Aliphatic Amines to α, β-Ethylenic Compounds
Synthetic Communications (2006)
Meshram HM, et al.