MilliporeSigma
  • Comparative molluscicidal action of extract of Ginko biloba sarcotesta, arecoline and niclosamide on snail hosts of Schistosoma japonicum
Comparative molluscicidal action of extract of <I>Ginko biloba</I> sarcotesta, arecoline and niclosamide on snail hosts of <I>Schistosoma japonicum</I>
Pesticide Biochemistry and Physiology (2007)
Chen, S., et al.