MilliporeSigma
  • OptGraft: A computational procedure for transferring a binding site onto an existing protein scaffold
OptGraft: A computational procedure for transferring a binding site onto an existing protein scaffold
Protein Science (2008)
Fazelinia, H., et al.