MilliporeSigma
15-lipoxygenase-1 in colorectal cancer: a review.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2009-09-16)
S Bhattacharya, G Mathew, D G Jayne, S Pelengaris, M Khan
ABSTRACT
Enzymes involved in the oxidative metabolism of n-6 polyunsaturated fatty acids, like lipoxygenase (LOX) and cyclooxygenase (COX), are significant in the pathogenesis of colorectal cancer. Of these enzymes, 15-LOX-1 is expressed in colon. Aim of this article is to describe the role and regulation of 15-LOX-1 in colorectal cancer and highlight its importance in cancer therapeutics. For our electronic literature research in PubMed and MEDLINE, key words related to 15-LOX-1 and colorectal cancer were used to find articles for this review. From the evidences, we believe that 15-LOX-1 has anti-carcinogenic effects in colorectal cancer, dependent or independent of its metabolites, and is manifested through downstream pathways involving cGMP, PPAR, p53, p21 and NAG-1, increasing apoptosis and decreasing proliferation in cancer cells. Regulation of 15-LOX-1 expression is achieved at transcription level by global histone acetylation and may also be dependent on GATA-6, IL-4 and IL-13. Positive relationship exists between 15-LOX-1 and survival in colorectal cancer. Evidences strongly support that therapeutic modulation of 15-LOX-1 may be a key to the treatment of colorectal cancer. However, it is still undecided whether the up-regulation of 15-LOX-1 alone can be sufficient to treat colorectal cancer and further studies are awaited.