MilliporeSigma
  • Rotational spectra and computational analysis of two conformers of leucinamide.
Rotational spectra and computational analysis of two conformers of leucinamide.
The journal of physical chemistry. A (2011-05-12)
Andrew R Conrad, Heather L Seedhouse, Richard J Lavrich, Michael J Tubergen
ABSTRACT
Rotational spectra were recorded for two isotopic species of two conformers of the amide derivative of leucine in the range of 10.5-21 GHz and fit to a rigid rotor Hamiltonian. Ab initio calculations at the MP2/6-311++G(d,p) level identified the low energy conformations with different side chain configurations; the rotational spectra were assigned to the two lowest energy ab initio structures. We recorded 16 a- and b-type rotational transitions for conformer 1; the rotational constants of the normal species are A = 2275.6(2), B = 1033.37(2) and C = 911.71(5) MHz. We recorded 23 a- and b-type rotational transitions for conformer 2; the rotational constants of the normal species are A = 2752.775(8), B = 843.502(1) and C = 796.721(1) MHz. The rotational spectra of the (15)N(amide) isotopomer of each conformer were recorded and the atomic coordinates of the amide nitrogen were determined by Kraitchman's method of isotopic substitution. The experimentally observed structures are significantly different from the crystal structures of leucinamide and the gas-phase structures of leucine, and a natural bond orbital analysis revealed the donor-acceptor interactions governing side chain configuration.
Sigma-Aldrich
L-Leucinamide hydrochloride, 99%