MilliporeSigma
  • The MFF-SIRT1/3 axis, regulated by miR-340-5p, restores mitochondrial homeostasis of hypoxia-induced pulmonary artery smooth muscle cells.
The MFF-SIRT1/3 axis, regulated by miR-340-5p, restores mitochondrial homeostasis of hypoxia-induced pulmonary artery smooth muscle cells.
Laboratory investigation; a journal of technical methods and pathology (2022-01-20)
Chun-Xia Huang, Zhi-Xin Jiang, Da-Yong Du, Zhi-Min Zhang, Yang Liu, Yun-Tian Li
ABSTRACT
Mitochondrial dynamics and quality control play a central role in the maintenance of the proliferation-apoptosis balance, which is closely related to the progression of pulmonary arterial hypertension (PAH). However, the exact mechanism of this balance remains unknown. Pulmonary artery smooth muscle cells (PASMCs) were cultured in hypoxia condition for constructing a PAH model in vitro. The expression of genes and proteins were determined by qRT-PCR and western bolt assays. Cell proliferation-apoptosis balance were tested by MTT, EdU and TUNEL assays. The mitochondrial functions were assessed by flow cytometry, JC-1, Mito tracker red staining, and corresponding kits. Besides, the molecular interaction was validated by dual-luciferase reporter assay. MFF was overexpressed in hypoxia-treated PAMSCs. Knockdown of MFF significantly repressed the excessive proliferation but enhanced cell apoptosis in hypoxia-treated PAMSCs. Moreover, MFF silencing improved mitochondrial function of hypoxia-treated PAMSCs by increasing ATP production and decreasing ROS release and mitochondrial fission. Mechanistically, MFF was a directly target of miR-340-5p, and could negatively regulate SIRT1/3 expression. Subsequently, functional rescue assays showed that the biological effects of MFF in hypoxia-treated PAMSCs were negatively regulated by miR-340-5p and depended on the regulation on SIRT1/3 pathway. These results provided evidences that miR-340-5p regulated MFF-SIRT1/3 axis to improve mitochondrial homeostasis and proliferation-apoptosis imbalance of hypoxia-treated PAMSCs, which provided a theoretical basis for the prevention and treatment of PAH.