MilliporeSigma
Chemical constituents of Melicope ptelefolia.
Natural product communications (2011-04-13)
Khozirah Shaari, Seema Zareen, Muhammad Nadeem Akhtar, Nordin Hj Lajis
ABSTRACT
Phytochemical investigations on the methanolic extract of Melicope ptelefolia Champ ex Benth. resulted in the isolation of three new compounds, identified as 3beta-stigmast-5-en-3-ol butyl tridecanedioate (melicoester) (1), (2Z, 6Z, 10Z, 14Z, 18Z, 22Z, 26E)-3', 7', 11', 15', 19', 23', 27', 31'-octamethyldotriaconta-2, 6, 10, 14, 18, 22, 26, 30-octadecanoate (melicopeprenoate) (2) and p-O-geranyl-7"-acetoxy coumaric acid (3). The compounds were isolated along with twenty-one other known compounds, lupeol (4), oleanolic acid (5), kokusaginine (6) genistein (7), p-O-geranyl coumaric acid (8), 4-stigmasten-3-one (9), 3beta-hydroxystigma-5-en-7-one (10) cis-phytyl palmitate (11), dodecane, dodecan-1-ol, ceryl alcohol, hentriacontanoic acid, eicosane, n-amyl alcohol, caprylic alcohol, octatriacontane, nonatriacontane, hexatriencontan-1-ol, methyl octacosanoate, beta-sitosterol, beta-sitosterol glucoside. Structures of all the compounds were established on the basis of MS and 1D and 2D NMR spectral data, as well as comparison with reported data.