MilliporeSigma
  • Synthesis, QSAR and anti-HIV activity of new 5-benzylthio-1,3,4-oxadiazoles derived from α-amino acids.
Synthesis, QSAR and anti-HIV activity of new 5-benzylthio-1,3,4-oxadiazoles derived from α-amino acids.
Journal of enzyme inhibition and medicinal chemistry (2011-01-22)
Tayyaba Syed, Tashfeen Akhtar, Najim A Al-Masoudi, Peter G Jones, Shahid Hameed
ABSTRACT
2-(1-[(4-Chloro/methylphenylsulfonylamino)alkyl]-5-thioxo-4,5-dihydro-1,3,4-oxadiazoles (4a-e) were synthesized, in four steps, via the sulfonyl derivatives of l-amino acids (l-alanine, l-methionine and l-phenylalanine) 1a-e, the esters 2a-e, the hydrazides 3a-e and finally the cyclization to 4a-e. Alkylation of 4a-e with 1.0 mole eq. of substituted benzyl halides furnished S-benzyl derivatives 5a-t, while 1.1 mole eq. yielded major 5a-t and minor amount of 6a-d. Alternatively, treatment of 4a-e with 2.0 mole eq. of substituted benzyl halides furnished 6a-d only. The structures of 5b and 5l were further confirmed by single crystal X-ray analysis. Compounds 5a-t and 6a-d showed no selective inhibition against HIV-1 and HIV-2 replication in MT-4 cells. However, 5f and 5j-5q exhibited some inhibitory activity against both types with EC(50) values (>11.50 - >13.00 µg/mL). These results suggest that the structural modifications of these compounds might lead to the development of new antiviral agents. The quantum structure-activity relationship of these novel structural congeners is discussed.