MilliporeSigma
Biological activities of leupeptins.
The Journal of antibiotics (1969-11-01)
T Aoyagi, S Miyata, M Nanbo, F Kojima, M Matsuzaki
Sigma-Aldrich
Leupeptin
Sigma-Aldrich
Leupeptin, >4000 U/mg