MilliporeSigma
  • Effects of Etanercept on TNF-α Inhibition in Rats with Adenine-Induced Chronic Kidney Disease.
Effects of Etanercept on TNF-α Inhibition in Rats with Adenine-Induced Chronic Kidney Disease.
BioMed research international (2022-06-02)
Edgar Mendieta-Condado, Elda Cristina Villaseñor-Tapia, Francisco Javier Gálvez-Gastelum, Irinea Yáñez-Sánchez, Oscar Pizano-Martínez, Alejandro Canales-Aguirre, Ana Laura Márquez-Aguirre
ABSTRACT
Chronic kidney disease (CKD) constitutes a chronic inflammatory state associated with an increase in inflammatory mediators and profibrotic molecules such as tumor necrosis factor-α (TNF-α). Etanercept (ETA) is a TNF inhibitor widely used in treatment of autoimmune inflammatory diseases. However, the effects of TNF-α inhibition in the establishment of CKD have not been fully elucidated. We evaluate the effects of TNF inhibition by ETA in adenine- (Ad-) induced CKD in rats. Rats were divided into three groups: control, renal injury model, and model plus ETA (2 mg/kg, 3 times per week (w); sc). Renal injury was induced by Ad administration (100 mg/kg, daily for 2 or 4 w; orogastric). Serum TNF-α levels and biochemical parameters for renal function were evaluated. Histopathological changes in the kidney were assessed using H&E and Masson's trichrome staining and also immunostaining for tubular cells. Ad administration produced a renal functional decline, tubular atrophy, interstitial inflammation, and fibrosis for 2 w, followed by renal anemia, several renal dysfunctions, tubular atrophy, and fibrosis at 4 w. A significant increase in serum TNF-α levels was observed from 2 w of Ad administration and remained elevated up to 4 w. Treatment with ETA partially reduced kidney damage but was very effective to blocking serum TNF-α. Although inhibition of TNF by ETA was very effective in reducing serum TNF-α, this strategy was partially effective in preventing Ad-induced CKD.
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant