MilliporeSigma
  • Structural development of a type-1 ryanodine receptor (RyR1) Ca2+-release channel inhibitor guided by endoplasmic reticulum Ca2+ assay.
Structural development of a type-1 ryanodine receptor (RyR1) Ca2+-release channel inhibitor guided by endoplasmic reticulum Ca2+ assay.
European journal of medicinal chemistry (2019-07-13)
Shuichi Mori, Hiroto Iinuma, Noriaki Manaka, Mari Ishigami-Yuasa, Takashi Murayama, Yoshiaki Nishijima, Akiko Sakurai, Ryota Arai, Nagomi Kurebayashi, Takashi Sakurai, Hiroyuki Kagechika
ABSTRACT
Type-1 ryanodine receptor (RyR1) is a calcium-release channel localized on sarcoplasmic reticulum (SR) of the skeletal muscle, and mediates muscle contraction by releasing Ca2+ from the SR. Genetic mutations of RyR1 are associated with skeletal muscle diseases such as malignant hyperthermia and central core diseases, in which over-activation of RyR1 causes leakage of Ca2+ from the SR. We recently developed an efficient high-throughput screening system based on the measurement of Ca2+ in endoplasmic reticulum, and used it to identify oxolinic acid (1) as a novel RyR1 channel inhibitor. Here, we designed and synthesized a series of quinolone derivatives based on 1 as a lead compound. Derivatives bearing a long alkyl chain at the nitrogen atom of the quinolone ring and having a suitable substituent at the 7-position of quinolone exhibited potent RyR1 channel-inhibitory activity. Among the synthesized compounds, 14h showed more potent activity than dantrolene, a known RyR1 inhibitor, and exhibited high RyR1 selectivity over RyR2 and RyR3. These compounds may be promising leads for clinically applicable RyR1 channel inhibitors.
Sigma-Aldrich
RyR1-selective inhibitor 1, ≥98% (HPLC)